missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyacetone 80.0+%, TCI America™

Catalog No. H038825ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H038825ML 25 mL
H0388100ML 100 mL
H0388500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H038825ML Supplier TCI America Supplier No. H038825ML

Chemical Identifiers

CAS 116-09-6
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00004669
InChI Key XLSMFKSTNGKWQX-UHFFFAOYSA-N
Synonym hydroxyacetone, acetol, acetone alcohol, 1-hydroxy-2-propanone, 2-propanone, 1-hydroxy, 1-hydroxyacetone, methanol, acetyl, acetylmethanol, acetylcarbinol, 2-oxopropanol
PubChem CID 8299
ChEBI CHEBI:27957
IUPAC Name 1-hydroxypropan-2-one
SMILES CC(=O)CO

Specifications

Color Yellow
Quantity 25 mL
UN Number 1993
Formula Weight 74.08
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Hydroxyacetone
EINECSNumber (2)-3205
RTECSNumber UC2800000
TSCA Yes
Recommended Storage Refrigerator