missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[Hydroxy(tosyloxy)iodo]benzene 97.0+%, TCI America™
Supplier: TCI America P101525G
Specifications
| [Hydroxy(tosyloxy)iodo]benzene | |
| 138°C | |
| C13H13IO4S | |
| 25 g | |
| LRIUKPUCKCECPT-UHFFFAOYSA-N | |
| [hydroxy(phenyl)-$l^{3}-iodanyl] 4-methylbenzenesulfonate | |
| 325434 | |
| ≥97.0% (T) |
| 27126-76-7 | |
| White-Yellow | |
| MFCD00011547 | |
| hydroxy tosyloxy iodo benzene, htib, koser's reagent, phenyliodosohydroxy tosylate, koser reagent, unii-2eoa97b14a, hydroxy p-tolylsulfonyl oxy iodo benzene, hydroxy 4-methylbenzenesulfonato-o phenyliodine, iodosobenzene-i-mono-p-toluenesulfonate, hydroxy phenyl 4-methylbenzenesulfonoperoxoyl iodide | |
| CC1=CC=C(C=C1)S(=O)(=O)OI(C2=CC=CC=C2)O | |
| 392.207 | |
| 392.21 | |
| Crystalline Powder |
Chemical Identifiers
| 27126-76-7 | |
| 392.207 | |
| LRIUKPUCKCECPT-UHFFFAOYSA-N | |
| 325434 | |
| CC1=CC=C(C=C1)S(=O)(=O)OI(C2=CC=CC=C2)O |
| C13H13IO4S | |
| MFCD00011547 | |
| hydroxy tosyloxy iodo benzene, htib, koser's reagent, phenyliodosohydroxy tosylate, koser reagent, unii-2eoa97b14a, hydroxy p-tolylsulfonyl oxy iodo benzene, hydroxy 4-methylbenzenesulfonato-o phenyliodine, iodosobenzene-i-mono-p-toluenesulfonate, hydroxy phenyl 4-methylbenzenesulfonoperoxoyl iodide | |
| [hydroxy(phenyl)-$l^{3}-iodanyl] 4-methylbenzenesulfonate |
Safety and Handling
TSCA : No