missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxy naphthol blue disodium salt, For ACS analysis, MP Biomedicals™
$65.50
Chemical Identifiers
| CAS | 165660-27-5 |
|---|---|
| Molecular Formula | C20H14N2Na2O11S3 |
| Molecular Weight (g/mol) | 600.495 |
| InChI Key | LXJHFXHZWYBBKJ-LUUCHEBKSA-N |
| Synonym | `-(2-naphtholazo-3, 6-disulfonic acid)-2-naphthol-4-sulfonic acid trisodium salt, calcium indicator on sodium chloride |
| PubChem CID | 131849366 |
| IUPAC Name | 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonic acid;sodium |
| SMILES | C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN15252910
|
N/A | 10 g |
Each for $65.50
|
|
|||||
Chemical Identifiers
| 165660-27-5 | |
| 600.495 | |
| `-(2-naphtholazo-3, 6-disulfonic acid)-2-naphthol-4-sulfonic acid trisodium salt, calcium indicator on sodium chloride | |
| 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonic acid;sodium |
| C20H14N2Na2O11S3 | |
| LXJHFXHZWYBBKJ-LUUCHEBKSA-N | |
| 131849366 | |
| C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na] |
Specifications
| 165660-27-5 | |
| `-(2-naphtholazo-3, 6-disulfonic acid)-2-naphthol-4-sulfonic acid trisodium salt, calcium indicator on sodium chloride | |
| C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na] | |
| 600.495 | |
| 598.5 | |
| Powder |
| C20H14N2Na2O11S3 | |
| LXJHFXHZWYBBKJ-LUUCHEBKSA-N | |
| 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonic acid;sodium | |
| 131849366 | |
| Black to Blue | |
| Hydroxy naphthol blue disodium salt, For ACS analysis |