missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxy Naphthol Blue, ACS Reagent, Reagents
Hydroxy Naphthol Blue, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
$253.11 - $570.64
Specifications
| Chemical Name or Material | Hydroxy Naphthol Blue, ACS Reagent |
|---|---|
| Boiling Point | 1413°C |
| CAS | 165660-27-5 |
| Molecular Formula | C20H12N2Na2O11S3 |
| Synonym | 1-(2-Naphtholazo-3,6-disulfonic Acid)-2-naphthol-4-sulfonic Acid Disodium Salt |
Specifications
| Hydroxy Naphthol Blue, ACS Reagent | |
| 165660-27-5 | |
| 1-(2-Naphtholazo-3,6-disulfonic Acid)-2-naphthol-4-sulfonic Acid Disodium Salt | |
| ACS | |
| 2.17 | |
| Solid |
| 1413°C | |
| C20H12N2Na2O11S3 | |
| 620.47 | |
| Glass Bottle | |
| Dark Blue |