missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[Hydroxy(methanesulfonyloxy)iodo]benzene 98.0+%, TCI America™
$141.67 - $697.16
Chemical Identifiers
| CAS | 105551-42-6 |
|---|---|
| Molecular Formula | C7H9IO4S |
| Molecular Weight (g/mol) | 316.11 |
| MDL Number | MFCD01321184 |
| InChI Key | WSUYPCBSGVAXNY-UHFFFAOYSA-N |
| Synonym | [Hydroxy(mesyloxy)iodo]benzene, Phenyliodosohydroxy Methanesulfonate |
| PubChem CID | 11141609 |
| IUPAC Name | [hydroxy(phenyl)-$l^{3}-iodanyl] methanesulfonate |
| SMILES | CS(=O)(=O)O[I](O)C1=CC=CC=C1 |
Chemical Identifiers
| 105551-42-6 | |
| 316.11 | |
| WSUYPCBSGVAXNY-UHFFFAOYSA-N | |
| 11141609 | |
| CS(=O)(=O)O[I](O)C1=CC=CC=C1 |
| C7H9IO4S | |
| MFCD01321184 | |
| [Hydroxy(mesyloxy)iodo]benzene, Phenyliodosohydroxy Methanesulfonate | |
| [hydroxy(phenyl)-$l^{3}-iodanyl] methanesulfonate |
Specifications
| 105551-42-6 | |
| White-Yellow | |
| MFCD01321184 | |
| [Hydroxy(mesyloxy)iodo]benzene, Phenyliodosohydroxy Methanesulfonate | |
| CS(=O)(=O)O[I](O)C1=CC=CC=C1 | |
| 316.11 | |
| 316.11 | |
| Crystalline Powder |
| 122°C | |
| C7H9IO4S | |
| 5 g | |
| WSUYPCBSGVAXNY-UHFFFAOYSA-N | |
| [hydroxy(phenyl)-$l^{3}-iodanyl] methanesulfonate | |
| 11141609 | |
| ≥98.0% (T) | |
| [Hydroxy(methanesulfonyloxy)iodo]benzene |
Safety and Handling
TSCA : No