missing translation for 'onlineSavingsMsg'
Learn More

[Hydroxy(methanesulfonyloxy)iodo]benzene 98.0+%, TCI America™

Catalog No. P12985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12985G 5 g
P129825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12985G Supplier TCI America Supplier No. P12985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105551-42-6
Molecular Formula C7H9IO4S
Molecular Weight (g/mol) 316.11
MDL Number MFCD01321184
InChI Key WSUYPCBSGVAXNY-UHFFFAOYSA-N
Synonym [Hydroxy(mesyloxy)iodo]benzene, Phenyliodosohydroxy Methanesulfonate
PubChem CID 11141609
IUPAC Name [hydroxy(phenyl)-$l^{3}-iodanyl] methanesulfonate
SMILES CS(=O)(=O)O[I](O)C1=CC=CC=C1

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 5 g
Formula Weight 316.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [Hydroxy(methanesulfonyloxy)iodo]benzene