Learn More
(Hydroxy-2-naphthylmethyl)phosphonic acid, 98%, Thermo Scientific Chemicals
A selective insulin receptor tyrosine kinase inhibitor
Supplier: Thermo Scientific Chemicals J63913MB
Description
A selective insulin receptor tyrosine kinase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (Hydroxy-2-naphthylmethyl)phosphonic acid | |
| White | |
| MFCD00674174 | |
| hnmpa, hydroxy-2-naphthalenylmethyl phosphonic acid, hydroxy 2-naphthyl methanephosphonic acid, hydroxy naphthalen-2-yl methylphosphonic acid, hydroxy naphthalen-2-yl methyl phosphonic acid, phosphonic acid, p-hydroxy-2-naphthalenylmethyl, oh-name-pa, acmc-1cglf, biomolki_000021, biomolki2_000010 | |
| C1=CC=C2C=C(C=CC2=C1)C(O)P(=O)(O)O | |
| 238.179 | |
| 238.18 | |
| Solid |
| 132541-52-7 | |
| C11H11O4P | |
| 25 mg | |
| AMJJLDJPDLKNJA-UHFFFAOYSA-N | |
| [hydroxy(naphthalen-2-yl)methyl]phosphonic acid | |
| 129317 | |
| 98% |
Chemical Identifiers
| 132541-52-7 | |
| 238.179 | |
| AMJJLDJPDLKNJA-UHFFFAOYSA-N | |
| 129317 | |
| C1=CC=C2C=C(C=CC2=C1)C(O)P(=O)(O)O |
| C11H11O4P | |
| MFCD00674174 | |
| hnmpa, hydroxy-2-naphthalenylmethyl phosphonic acid, hydroxy 2-naphthyl methanephosphonic acid, hydroxy naphthalen-2-yl methylphosphonic acid, hydroxy naphthalen-2-yl methyl phosphonic acid, phosphonic acid, p-hydroxy-2-naphthalenylmethyl, oh-name-pa, acmc-1cglf, biomolki_000021, biomolki2_000010 | |
| [hydroxy(naphthalen-2-yl)methyl]phosphonic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only