missing translation for 'onlineSavingsMsg'
Learn More

(Hydroxy-2-naphthylmethyl)phosphonic acid, 98%, Thermo Scientific Chemicals

Catalog No. p-7042830
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63913LB0 5 mg
AAJ63913MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63913LB0 Supplier Thermo Scientific Chemicals Supplier No. J63913LB0
Only null left

A selective insulin receptor tyrosine kinase inhibitor

A selective insulin receptor tyrosine kinase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132541-52-7
Molecular Formula C11H11O4P
Molecular Weight (g/mol) 238.179
MDL Number MFCD00674174
InChI Key AMJJLDJPDLKNJA-UHFFFAOYSA-N
Synonym hnmpa, hydroxy-2-naphthalenylmethyl phosphonic acid, hydroxy 2-naphthyl methanephosphonic acid, hydroxy naphthalen-2-yl methylphosphonic acid, hydroxy naphthalen-2-yl methyl phosphonic acid, phosphonic acid, p-hydroxy-2-naphthalenylmethyl, oh-name-pa, acmc-1cglf, biomolki_000021, biomolki2_000010
PubChem CID 129317
IUPAC Name [hydroxy(naphthalen-2-yl)methyl]phosphonic acid
SMILES C1=CC=C2C=C(C=CC2=C1)C(O)P(=O)(O)O

Specifications

Color White
Quantity 5 mg
Formula Weight 238.18
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (Hydroxy-2-naphthylmethyl)phosphonic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only