missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Water Standard 10.0 mg/g, Ricca Chemical
Description
- Reliable, Safe, Pyridine-Free
- Stable Formulations and Comprehensive Chemical Compatibility
- 5 Year Shelf Life for Most Reagents
- Technical Support Every Step of the Way
- Tight Specifications for More Accurate Results and Low Lot-to-Lot Variability
- Serving Scientists Successfully for Over 45 Years
Specifications
Specifications
| Chemical Name or Material | Water Standard |
| CAS Min % | 40, 5, 1 |
| CAS Max % | 40, 50, 5 |
| CAS | 100-41-4, 7732-18-5, 1330-20-7, 71-36-3, 75-12-7 |
| Quantity | 10 x 5 mL |
| Packaging | Glass Ampule |
| Grade | Karl Fischer |
| Concentration or Composition (by Analyte or Components) | 10 mg/g |