missing translation for 'onlineSavingsMsg'
Learn More

HydroSpec™ Water Standard 0.10 mg/g, Ricca Chemical

Catalog No. K410000105R
Change view
Click to view available options
Quantity:
10 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K410000105R 10 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K410000105R Supplier Ricca Chemical Company Supplier No. RK41000010X5R
Only null left

Achieve high performance and reliable results. Hydrospec™ supports a wide range of sample types that help you produce the fast, reliable, and reproducible results you are looking for.

  • Reliable, Safe, Pyridine-Free
  • Stable Formulations and Comprehensive Chemical Compatibility
  • 5 Year Shelf Life for Most Reagents
  • Technical Support Every Step of the Way
  • Tight Specifications for More Accurate Results and Low Lot-to-Lot Variability
  • Serving Scientists Successfully for Over 45 Years

Specifications

Chemical Name or Material Water Standard
CAS Min % 0.01, 18
CAS Max % 81.99, 0.01
CAS 7732-18-5, 100-41-4, 1330-20-7, 100-41-4
Quantity 10 x 5 mL
Packaging Glass Ampule
Grade Karl Fischer
Concentration or Composition (by Analyte or Components) 0.1 mg/g