missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Titrant T5, Ricca Chemical
Spécifications
Spécifications
| CAS Min % | 85, 5 |
| CAS | 67-56-1, 7553-56-2 |
| Physical Form | Liquid |
| Quantity | 1 L |
| CAS Max % | 15 |
| Packaging | Amber Glass Bottle |
| UN Number | UN1230 |
| Specific Gravity | 0.89 |