missing translation for 'onlineSavingsMsg'
Learn More

HydroSpec™ Titrant T2, Ricca Chemical

Numéro de catalogue. RK2500001C
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
RK2500001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. RK2500001C Fournisseur Ricca Chemical Company Code fournisseur RK2500001C
Il en reste null

Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.

Spécifications

CAS Min % 85, 1
CAS 67-56-1, 7553-56-2
Physical Form Liquid
Quantity 1 L
CAS Max % 5
Packaging Amber Glass Bottle
UN Number UN1230
Specific Gravity 0.89