missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Solvent, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Solvent |
| CAS Min % | 5, 10 |
| CAS Max % | 10, 95 |
| CAS | 7446-09-5, 288-32-4, 67-56-1 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Packaging | Amber Glass Bottle |
| UN Number | UN1230 |
| Specific Gravity | 0.91 |
| Grade | Karl Fischer |