missing translation for 'onlineSavingsMsg'
Learn More

HydroSpec™ Solvent CX, Ricca Chemical

Catalog No. RK3200001C
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RK3200001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RK3200001C Supplier Ricca Chemical Company Supplier No. RK3200001C
Only null left

Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.

Specifications

Chemical Name or Material Solvent CX
CAS Min % 5, 10
CAS Max % 40, 10, 10
CAS 1330-20-7, 288-32-4, 67-66-3, 67-56-1, 7446-09-5
Physical Form Liquid
Quantity 1 L
Packaging Amber Glass Bottle
UN Number UN1992
Specific Gravity 1.04
Grade Karl Fischer