missing translation for 'onlineSavingsMsg'
Learn More

HydroSpec™ Solvent C, Ricca Chemical

Numéro de catalogue. RK3300001C
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RK3300001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RK3300001C Supplier Ricca Chemical Company Supplier No. RK3300001C
Only null left

Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.

Specifications

Chemical Name or Material Solvent C
CAS Min % 5, 20
CAS Max % 10
CAS 7446-09-5, 67-56-1, 67-66-3, 288-32-4
Physical Form Liquid
Quantity 1 L
Packaging Amber Glass Bottle
UN Number UN1992
Specific Gravity 1.04
Grade Karl Fischer