missing translation for 'onlineSavingsMsg'
Learn More

HydroSpec™ Methanol Accelerate, Ricca Chemical

Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.

$240.46 - $240.46

Specifications

Chemical Name or Material Methanol Accelerate
CAS Min % 85,5
Physical Form Liquid
CAS 67-56-1
Molecular Formula CH4O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RK3000001C
View Documents
Ricca Chemical Company
RK3000001C
1 L N/A
Only null left
 
Description

Description

Specifications

Specifications

Methanol Accelerate
Liquid
CH4O
Amber Glass Bottle
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
100%
85,5
67-56-1
MFCD00004595
0.91
CO
32.04
Karl Fischer
Videos
SDS
Documents

Documents