missing translation for 'onlineSavingsMsg'
Learn More

HydroSpec™ Methanol Accelerate, Ricca Chemical

Catalog No. RK3000001C
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RK3000001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RK3000001C Supplier Ricca Chemical Company Supplier No. RK3000001C
Only null left

Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.

Specifications

Chemical Name or Material Methanol Accelerate
CAS Min % 85, 5
Physical Form Liquid
CAS 67-56-1
Quantity 1 L
CAS Max % 95
Molecular Formula CH4O
MDL Number MFCD00004595
Packaging Amber Glass Bottle
Specific Gravity 0.91
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Percent Purity 100%
Grade Karl Fischer
Show More Show Less