missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Coulometric AH, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Coulometric AH |
| CAS Min % | 5, 30 |
| CAS Max % | 25, 10 |
| CAS | 7446-09-5, 112-30-1, 67-56-1, 288-32-4 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Packaging | Amber Glass Bottle |
| UN Number | UN1230 |
| Specific Gravity | 0.93 |
| Grade | Karl Fischer |