missing translation for 'onlineSavingsMsg'
Learn More

HydroSpec™ Coulometric AG, Ricca Chemical

Catalog No. RK100000500
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RK100000500 500 mL
RK1000001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RK100000500 Supplier Ricca Chemical Company Supplier No. RK100000500C
Only null left

Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.

Specifications

Chemical Name or Material Coulometric AG
CAS Min % 10
CAS Max % 30
CAS 288-32-4, 68007-08-9, 7446-09-5, 67-56-1, 111-42-2
Physical Form Liquid
Quantity 500 mL
Packaging Amber Glass Bottle
UN Number UN1230
Specific Gravity 0.93
Grade Karl Fischer