missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Composit T5, Ricca Chemical
Specifications
Specifications
| CAS Min % | 5, 10 |
| CAS Max % | 20, 15 |
| CAS | 7446-09-5, 288-32-4, 111-90-0, 7553-56-2 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Packaging | Amber Glass Bottle |
| UN Number | UN3082 |
| Specific Gravity | 1.1 |