missing translation for 'onlineSavingsMsg'
Learn More

HydroSpec™ Composit K5, Ricca Chemical

Catalog No. RK2200001C
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RK2200001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RK2200001C Supplier Ricca Chemical Company Supplier No. RK2200001C
Only null left

Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.

Specifications

CAS Min % 5, 10
CAS Max % 20, 15
CAS 7446-09-5, 288-32-4, 111-90-0, 7553-56-2
Physical Form Liquid
Quantity 1 L
Packaging Amber Glass Bottle
UN Number UN3082
Specific Gravity 1.1