missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Acid Buffer, Ricca Chemical
Description
- Reliable, Safe, Pyridine-Free
- Available in 500 mL and 1 L sizes
- 1 L Bottles have the Industry Standard GL45 Neck Finish for Convenient Connection
- Stable Formulations and Comprehensive Chemical Compatibility
- 5 Year Shelf Life for Most Reagents
- Technical Support Every Step of the Way
- Tight Specifications for More Accurate Results and Low Lot-to-Lot Variability
- Serving Scientists Successfully for Over 45 Years
Specifications
Specifications
| Chemical Name or Material | Acid Buffer |
| CAS Min % | 90, 10 |
| CAS | 64-56-1, 288-32-4, 67-56-1, 288-32-4 |
| CAS Max % | 90, 10 |
| Quantity | 1 L |
| Packaging | Amber Glass Bottle |
| Grade | Karl Fischer |