missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinonesulfonic acid potassium salt, 98+%

Catalog No. AAL0840636
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0840636 500 g
AAL0840622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0840636 Supplier Thermo Scientific Chemicals Supplier No. L0840636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21799-87-1 | C6H5KO5S | 228.259 g/mol

It is employed in the synthesis and properties of sulfonated polyarylene ether nitrile copolymers for PEM with high thermal stability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis and properties of sulfonated polyarylene ether nitrile copolymers for PEM with high thermal stability.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 21799-87-1
Molecular Formula C6H5KO5S
Molecular Weight (g/mol) 228.259
MDL Number MFCD00007475
InChI Key VKDSBABHIXQFKH-UHFFFAOYSA-M
Synonym potassium 2,5-dihydroxybenzenesulfonate, hydroquinonesulfonic acid potassium salt, potassium2,5-dihydroxybenzenesulfonate, unii-249e3f00ep, benzenesulfonic acid, 2,5-dihydroxy-, monopotassium salt, 2,5-dihydroxybenzenesulfonic acid potassium salt, potassium hydroquinonesulfonate, hydroquinone sulfonic acid potassium salt, hydroquinone sulfonic acid, potassium salt, benzenesulfonic acid, 2,5-dihydroxy-, potassium salt 1:1
PubChem CID 23672329
IUPAC Name potassium;2,5-dihydroxybenzenesulfonate
SMILES C1=CC(=C(C=C1O)S(=O)(=O)[O-])O.[K+]

Specifications

Melting Point ∽251°C (decomposition)
Quantity 500 g
Beilstein 5187027
Solubility Information Soluble in water.
Formula Weight 228.27
Percent Purity ≥98%
Chemical Name or Material Hydroquinonesulfonic acid potassium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 244-584-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only