missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone 99.0+%, TCI America™

Catalog No. p-7130392
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H018625G 25 g
H0186500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H018625G Supplier TCI America Supplier No. H018625G
Only null left

Chemical Identifiers

CAS 123-31-9
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00002339
InChI Key QIGBRXMKCJKVMJ-UHFFFAOYSA-N
Synonym hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol
PubChem CID 785
ChEBI CHEBI:17594
IUPAC Name benzene-1,4-diol
SMILES C1=CC(=CC=C1O)O

Specifications

Melting Point 173°C
Color White
Boiling Point 282°C
Quantity 25 g
UN Number 3077
Formula Weight 110.11
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hydroquinone
EINECSNumber (3)-0543
RTECSNumber MX3500000
TSCA Yes