missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone, Crystal, 98.5%, Spectrum™ Chemical

Catalog No. 18610968
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610968 125 g Poly Pail
18610971 500 g Poly Pail
18610969 2.5 kg Poly Pail
18610967 12 kg Poly Pail
18610970 50 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610968 Supplier Spectrum Chemical Mfg Cor Supplier No. H1080125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 123-31-9

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 125 g
Percent Purity 0.985
Grade Reagent
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.