missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone bis(2-hydroxyethyl)ether, 98+%

Catalog No. AC218220010
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC218220010 1 kg
AC218221000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC218220010 Supplier Thermo Scientific Chemicals Supplier No. 218220010
Only null left

CAS: 104-38-1 | C10H14O4 | 198.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-38-1
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00002861
InChI Key WTPYFJNYAMXZJG-UHFFFAOYSA-N
Synonym hydroquinone bis 2-hydroxyethyl ether, 1,4-bis 2-hydroxyethoxy benzene, vernatzer 30/10, 2,2'-1,4-phenylenebis oxy diethanol, hydroquinone di 2-hydroxyethyl ether, hydroquinone diethylol ether, 2,2'-phenylenedioxy diethanol, 2,2'-p-phenylenedioxy diethanol, 2,2'-p-phenylenedioxydiethanol, 2-4-2-hydroxyethoxy phenoxy ethanol
PubChem CID 66912
IUPAC Name 2-[4-(2-hydroxyethoxy)phenoxy]ethanol
SMILES C1=CC(=CC=C1OCCO)OCCO

Specifications

Melting Point 104.0°C to 107.0°C
Color White
Boiling Point 185.0°C to 200.0°C (0.3 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula C6H4(OCH2CH2OH)2
Quantity 1 kg
Beilstein 06, III, 4406
Solubility Information Solubility in water: practically insoluble. Other solubilities: ca 4g/100mL ethanol,ca 4g/100mL acetone
Formula Weight 198.22
Percent Purity 99+%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Hydroquinone bis(2-hydroxyethyl)ether, 99+%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 203-197-3
TSCA TSCA

RUO – Research Use Only