missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, technical, 35 wt% solution in water, stabilized, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 108.0°C |
| Melting Point | -33.0°C |
| Linear Formula | H2O2 |
| Merck Index | 15,4835 |
| Density | 1.1350g/mL |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol and sulfuric ether, insoluble in petroleum ether |
| Formula Weight | 34 |
| Grade | Technical |
| Packaging | Plastic Drum |
| Specific Gravity | 1.135 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May intensify fire; oxidizer. Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye damage. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immedia |
| EINECSNumber | 231-765- |
RUO – Research Use Only