missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen perOxide Solution, purum p.a., ≥35% (RT), Honeywell™
Purum p.a.,≥35% (RT)
$633.32 - $633.32
Chemical Identifiers
| CAS | 7722-84-1 |
|---|---|
| Molecular Formula | H2O2 |
| Molecular Weight (g/mol) | 34.014 |
| MDL Number | MFCD00011333 |
| InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
| Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
| PubChem CID | 784 |
| ChEBI | CHEBI:16240 |
| IUPAC Name | hydrogen peroxide |
| SMILES | OO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
6000805
|
Honeywell Inc
9529925L |
2.5 L |
N/A
|
N/A | |||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
| 7722-84-1 | |
| 34.014 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| 784 | |
| hydrogen peroxide |
| H2O2 | |
| MFCD00011333 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| CHEBI:16240 | |
| OO |
Specifications
| 7722-84-1 | |
| MFCD00011333 | |
| 3587191 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| OO | |
| 34.014 | |
| CHEBI:16240 | |
| ≥35% (RT) | |
| 2.5 L |
| H2O2 | |
| UN2014 | |
| 1.13g/mL at 20°C | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| hydrogen peroxide | |
| 784 | |
| 34.01g/mol | |
| 23.3 mmHg (30°C) | |
| Hydrogen peroxide solution |
Safety and Handling
P280-P305 + P351 + P338 + P310