missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide Solution AR 30%, Macron Fine Chemicals™ ™
macron
$11138.00 - $11138.00
Chemical Identifiers
| CAS | 7722-84-1 |
|---|---|
| Molecular Formula | H2O2 |
| Molecular Weight (g/mol) | 34.014 |
| InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
| Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
| PubChem CID | 784 |
| ChEBI | CHEBI:16240 |
| IUPAC Name | hydrogen peroxide |
| SMILES | OO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
MKV340500
|
Avantor
V34004 |
500 mL | Poly |
N/A
|
N/A | ||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
Chemical Identifiers
| 7722-84-1 | |
| 34.014 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| CHEBI:16240 | |
| OO |
| H2O2 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| 784 | |
| hydrogen peroxide |
Specifications
| 7722-84-1 | |
| H2O2 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| hydrogen peroxide | |
| 784 | |
| 34.01 | |
| 500 mL |
| 30% | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| OO | |
| 34.014 | |
| CHEBI:16240 | |
| Poly | |
| Hydrogen Peroxide Solution |