missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide Solution, ACS Reagent, Honeywell™
ACS Reagent, contains inhibitor, 30Â wt. % in H2O
$1637.21 - $1637.21
Chemical Identifiers
| CAS | 7722-84-1 |
|---|---|
| Molecular Formula | H2O2 |
| Molecular Weight (g/mol) | 34.014 |
| MDL Number | MFCD00011333 |
| InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
| Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
| PubChem CID | 784 |
| ChEBI | CHEBI:16240 |
| IUPAC Name | hydrogen peroxide |
| SMILES | OO |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
6000801
|
Honeywell Inc
2167634L |
4 L |
N/A
|
N/A | |||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
| 7722-84-1 | |
| 34.014 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| 784 | |
| hydrogen peroxide |
| H2O2 | |
| MFCD00011333 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| CHEBI:16240 | |
| OO |
Specifications
| Undesignated | |
| H2O2 | |
| 2014 | |
| 3587191 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| hydrogen peroxide | |
| 784 | |
| 34.01g/mol | |
| 23.3mmHg (30°C) | |
| 4 L |
| 7722-84-1 | |
| MFCD00011333 | |
| 30wt. % in H2O | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| OO | |
| 34.014 | |
| CHEBI:16240 | |
| ACS Reagent | |
| 1 (vs air) | |
| Hydrogen Peroxide Solution, Contains inhibitor |
Safety and Handling
P280-P305 + P351 + P338 + P310
H318-H412