missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, Certified, 3.0% (w/w) to 3.5% (w/w), LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 97, 3 |
| Physical Form | Liquid |
| Quantity | 4 L |
| Linear Formula | H2O2 |
| Solubility Information | Soluble in water |
| Formula Weight | 34.01 |
| Percent Purity | 3.0 to 3.5% (w/w) |
| Grade | Certified |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye damage. |
| Precautionary Statement | GHS P Statement Wear protective gloves, eye protection. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. If skin irritation occurs: Get medical attention. Take off contaminated clothing and wash before reuse. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. |
| Recommended Storage | Room Temperature |