missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide, for analysis, 35 wt.% solution in water, stabilized

Catalog No. p-4090151
Change view
Click to view available options
Quantity:
25 mL
2.5 L
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC202460250 25 mL Plastic bottle
AC202460025 2.5 L Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC202460250 Supplier Thermo Scientific Chemicals Supplier No. 202460250

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7722-84-1 | H2O2 | 34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Boiling Point 108.0°C
Melting Point -33.0°C
Color Yellow through Brown
Physical Form Powder
Quantity 25 mL
Linear Formula H2O2
Fieser 01,457; 02,216; 03,154; 04,253; 05,337; 06,286; 07,174; 08,247; 09,241; 10,201; 11,256; 12,242; 14,176; 15,166; 16,177; 17,145
Merck Index 15, 4835
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and sulfuric ether, insoluble in petroleum ether
Formula Weight 34
Percent Purity 34.5 to 37%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 1.135
Density 1.1350g/mL
Chemical Name or Material Hydrogen peroxide, For analysis, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immedia
EINECSNumber 231-765-

RUO – Research Use Only