Learn More
Hydrogen peroxide, ACS reagent, 30 wt% solution in water, nonstabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 411881000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
| Hydrogen peroxide | |
| 7722-84-1,7732-18-5 | |
| -33.0°C | |
| HOOH | |
| 01,457; 02,216; 03,154; 04,253; 05,337; 06,286; 07,174; 08,247; 09,241; 10,201; 11,256; 12,242; 14,176; 15,166; 16,177; 17,145 | |
| 15, 4835 | |
| Solubility in water: soluble. Other solubilities: soluble in ether, alcohol, insoluble in petroleum ether, decomposed by many organic solvents | |
| OO | |
| 34.014 | |
| CHEBI:16240 | |
| 29.0 to 32.0% | |
| Plastic bottle | |
| 1.11 | |
| 100 g |
| Undesignated | |
| 108.0°C | |
| H2O2 | |
| MFCD00011333 | |
| 1.1100g/mL | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| hydrogen peroxide | |
| 784 | |
| 34 | |
| ACS Reagent | |
| 0.002% max. | |
| >125°C | |
| Solution |
Chemical Identifiers
| 7722-84-1 | |
| 34.014 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| 784 | |
| hydrogen peroxide |
| H2O2 | |
| MFCD00011333 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| CHEBI:16240 | |
| OO |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 231-765-0
RTECSNumber : MX0890000
TSCA : TSCA
RUO – Research Use Only