Learn More
Hydrogen peroxide, ACS, 29-32% w/w aq. soln., stab.
Description
For bleaching and deodorizing textiles, wood, hair, etc; rocket propulsion; maturing agent in food; water and sewage treatment; disinfectant; laboratory reagentHydrogen peroxide is used as an oxidizing agent to prepare propylene oxide, dibenzoyl peroxide, lauryl peroxide, peracetic acid and meta-chloroperoxybenzoic acid. It is also involved in the production of inorganic chemicals like sodium perborate and sodium percarbonate, which are used as bleaching agents in detergents. It is an associated iron catalyst and utilized for waste-water treatment to remove organic impurities through an advanced oxidation process. It is also used as a laboratory disinfectant and as a topical antiseptic. In the cosmetic industry, it is used for bleaching human hair and tooth whitening by mixing with ammonium hydroxide and baking soda respectively. Monopropellant engines using hydrogen peroxide as the fuel are commonly used for air-independent underwater propulsion applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 107°C |
| Beilstein | 3587191 |
| Density | 1.11 g/cm3 |
| Solubility Information | Soluble in water,alcohol and ether. Insoluble in petroleumMiscible with water,ether,alcohol,organic solvents and carboxylic esters. Immiscible with hydrocarbons and petroleum ether. |
| Formula Weight | 34.02 |
| Grade | ACS |
| Refractive Index | 1.335 |
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Quantity | 50 mL |
| Show More |
Safety and Handling
| Hazard Category | H271-H302+H332-H314 |
| Precautionary Statement | P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P370+P378q-P501c |
| DOTInformation | Hazard Class: 5.1, Packing Group: II |
| EINECSNumber | 231-765-0 |
| RTECSNumber | MX0899000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only