missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide, ACS, 29-32% w/w aq. soln., stab.

Catalog No. AA33323AD
Change view
Click to view available options
Quantity:
50 mL
500 mL
4 x 500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33323AD 50 mL
AA33323AP 500 mL
AA33323AY 4 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA33323AD Supplier Thermo Scientific Chemicals Supplier No. 033323AD

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Bleaching and Deodorizing Textiles, Wood, Hair | CAS: 7722-84-1 | H2O2 | 34.014 g/mol

For bleaching and deodorizing textiles, wood, hair, etc; rocket propulsion; maturing agent in food; water and sewage treatment; disinfectant; laboratory reagentHydrogen peroxide is used as an oxidizing agent to prepare propylene oxide, dibenzoyl peroxide, lauryl peroxide, peracetic acid and meta-chloroperoxybenzoic acid. It is also involved in the production of inorganic chemicals like sodium perborate and sodium percarbonate, which are used as bleaching agents in detergents. It is an associated iron catalyst and utilized for waste-water treatment to remove organic impurities through an advanced oxidation process. It is also used as a laboratory disinfectant and as a topical antiseptic. In the cosmetic industry, it is used for bleaching human hair and tooth whitening by mixing with ammonium hydroxide and baking soda respectively. Monopropellant engines using hydrogen peroxide as the fuel are commonly used for air-independent underwater propulsion applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
Boiling Point 107°C
Beilstein 3587191
Density 1.11 g/cm3
Solubility Information Soluble in water,alcohol and ether. Insoluble in petroleumMiscible with water,ether,alcohol,organic solvents and carboxylic esters. Immiscible with hydrocarbons and petroleum ether.
Formula Weight 34.02
Grade ACS
Refractive Index 1.335
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Quantity 50 mL
Physical Form Liquid
Chemical Name or Material Hydrogen peroxide
Show More Show Less
Hazard Category H271-H302+H332-H314
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P370+P378q-P501c
DOTInformation Hazard Class: 5.1, Packing Group: II
EINECSNumber 231-765-0
RTECSNumber MX0899000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only