missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, 50%, Solution, Reagent, 48.0-51.0%, Spectrum™ Chemical

Catalog No. 18610959
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
200 L
Packaging:
Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610959 100 mL Poly Bottle
18610960 500 mL Poly Bottle
18610963 4 L Poly Bottle
18610958 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610959 Supplier Spectrum Chemical Mfg Cor Supplier No. H1077100ML

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Hydrogen Peroxide
CAS 7722-84-1, 7732-18-5
CAS Max % 50
Concentration 50%
Percent Purity 48 to 51%
Grade Reagent
Packaging Poly Bottle
pH 2
Quantity 100 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.