missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, 30% w/w, ACS Reagent , Reagents
Specifications
Specifications
| Color | Colorless |
| CAS | 7722-84-1 |
| Boiling Point | 107°C |
| Melting Point | -26°C |
| Molecular Formula | H2O2 |
| InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
| SMILES | OO |
| IUPAC Name | peroxol |
| Molecular Weight (g/mol) | 34.01 |
| Grade | ACS |
| Show More |