missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, 30% w/w, ACS Reagent , Reagents

Catalog No. 01337934
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337934 1 L
01337933 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337934 Supplier Labchem Inc Supplier No. C12146001AV
Only null left

Hydrogen Peroxide, 30% w/w, ACS Reagent , ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

Synonym Dihydrogen Dioxide, Oxydol

Specifications

Color Colorless
CAS 7722-84-1
Boiling Point 107°C
Melting Point -26°C
Molecular Formula H2O2
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILES OO
IUPAC Name peroxol
Molecular Weight (g/mol) 34.01
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.11
pH 1 to 4
Quantity 1 L
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide, 30% w/w, ACS Reagent
Show More Show Less