missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, 30% (w/w), ACS Reagent Grade, Ricca Chemical

$941.06 - $2,109.84

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
38217-32
SDS View Documents
Ricca Chemical Company
3821732
1 L Vented Poly Bottle N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
3821-71
SDS View Documents
Ricca Chemical Company
382171
4 L Vented Poly Bottle N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
38217-5
SDS View Documents
Ricca Chemical Company
382175
20 L Cubitainer N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7722-84-1
34.014
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
CHEBI:16240
OO
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
784
hydrogen peroxide

Specifications

Colorless
25.0,65.0
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
hydrogen peroxide
784
30%
30% (w/w)
3.3
Liquid
7722-84-1,7732-18-5
35.0,75.0
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
OO
34.014
CHEBI:16240
ACS Reagent
Vented Poly Bottle
1 L
Hydrogen Peroxide
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'