missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen peroxide 30%, Suprapur™, MilliporeSigma™
Supplier: Technidata 1072981000
Specifications
| Hydrogen peroxide | |
| Suprapur™ | |
| 30% | |
| H2;O2 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| hydrogen peroxide | |
| 784 | |
| Suprapur | |
| 18 hPa (20°C) | |
| Liquid |
| Colorless | |
| 7722-84-1 | |
| H2O2 | |
| 1.11g/cm3 (20°C) | |
| Soluble (20°C) | |
| OO | |
| 34.014 | |
| CHEBI:16240 | |
| Plastic Bottle | |
| 1 L |
Chemical Identifiers
| 7722-84-1 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| CHEBI:16240 |
| H2O2 | |
| 784 | |
| OO |
Safety and Handling
SignalWord: Danger
Recommended Storage : Store at +5°C to +30°C