missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Hydrogen Peroxide, 30% (Certified ACS), Fisher Chemical™
Manufacturer: Fisher Chemical H3254
Specifications
Hydrogen Peroxide, 30% | |
Vapor Pressure: 23mmHg at 30°C | |
29.0 to 32.0 % | |
H2O2 | |
1.110g/cm³ | |
albone, hioxyl, hydrogen dioxide, inhibine, interox, kastone, oxydol, perhydrol, peroxaan, superoxol | |
OO | |
34.014 | |
CHEBI:16240 | |
Liquid | |
Certified ACS | |
Pass test | |
0.1ppm max. | |
Poly Bottle | |
0.002 % max. | |
0.0006meq/g max. | |
Decomposes information: >125°C | |
Colorless | |
3.3 |
7722-84-1,7732-18-5 | |
5ppm max. | |
2ppm max. | |
MFCD00011333 | |
0.1ppm max. | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
34 | |
29.0 to 32.0% | |
1ppm max. | |
0.5ppm max. | |
2ppm max. | |
2ppm max. | |
5ppm max. | |
23mmHg at 30°C | |
108°C | |
-33°C | |
4L |
Chemical Identifiers
7722-84-1 | |
34.014 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
784 | |
hydrogen peroxide |
H2O2 | |
MFCD00011333 | |
albone, hioxyl, hydrogen dioxide, inhibine, interox, kastone, oxydol, perhydrol, peroxaan, superoxol | |
CHEBI:16240 | |
OO |
Safety and Handling

DANGER!
Emergency Overview
Causes severe eye irritation and possible burns. Irritating to skin. Harmful if swallowed.
NFPA
Health:3 Flammability:0 Instability:1
Signal Word: DANGER!
DOTInformation : DOT Class 5.1, : Oxidizer