missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide, 27% w/w aq. soln., stab.

Catalog No. p-7046129
Change view
Click to view available options
Quantity:
2 x 50 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13235BEX 2 x 50 mL
AAL13235AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL13235BEX Supplier Thermo Scientific Chemicals Supplier No. L13235BEX
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Boiling Point 107°C
Melting Point -26°C
UN Number UN2014
Beilstein 3587191
Concentration 27% w/w aq. Soln
Density 1.1
Merck Index 14,4798
Solubility Information Miscible with water,ether,alcohol,organic solvents and carboxylic esters. Immiscible with hydrocarbons and petroleum ether.
Formula Weight 34.02
Refractive Index 1.335
Quantity 2 x 50 mL
Chemical Name or Material Hydrogen peroxide, stabilized
Show More Show Less
Hazard Category H271-H302+H332-H314
Hazard Statement GHS H Statement
H272-H318-H302
May intensify fire; oxidizer.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: HYDROGEN PEROXIDE, AQUEOUS SOLUTIONS
EINECSNumber 231-765-0
RTECSNumber MX0899000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only