missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, 2% (w/w), Ricca Chemical
Supplier: Ricca Chemical Company R38180001B
| Quantity | 1 L |
|---|---|
| Packaging | Amber Poly Bottle |
Chemical Identifiers
| 7722-84-1 , 7732-18-5 | |
| 34.014 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| CHEBI:16240 | |
| OO |
| H2O2 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| 784 | |
| hydrogen peroxide |
Specifications
| Hydrogen Peroxide | |
| 97.0, 1.0 | |
| H2O2 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| hydrogen peroxide | |
| 784 | |
| 2% (w/w) Aqueous Solution | |
| Amber Poly Bottle |
Safety and Handling
ShelfLife : 12 months