missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide, 12% w/v

CAS: 7722-84-1 | H2O2 | 34.01 g/mol

$82.63 - $82.63

Chemical Identifiers

CAS 7722-84-1
Molecular Weight (g/mol) 34.01
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
IUPAC Name peroxol
SMILES OO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC464570010
SDS View Documents
Thermo Scientific Chemicals
464570010
1 L
Each for $82.63
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7722-84-1
MHAJPDPJQMAIIY-UHFFFAOYSA-N
OO
34.01
peroxol

Specifications

7722-84-1
1.0500 g/mL
H2O2
1.05
MHAJPDPJQMAIIY-UHFFFAOYSA-N
peroxol
34.01
100.0°C
1 L
H2O2
Solubility aq. soln.: soluble.
OO
34.01
Hydrogen peroxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement May intensify fire; oxidizer.

Causes serious eye damage.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Wash face, hands and any exposed skin thoroughly after handling.

Do not eat, drink or smoke when using this product.

GHS Signal Word: Danger

EINECSNumber : 231-765-0

SDS
Documents

Documents

RUO – Research Use Only