missing translation for 'onlineSavingsMsg'
Learn More

Gastec Corporation™ Hydrogen Cyanide Detector Tube

Catalog No. 17532384
Change view
Click to view available options
Detection Limits:
0.2 to 10 ppm
17 to 2400 ppm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Limits
17532384 0.2 to 10 ppm
17532383 17 to 2400 ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17532384 Supplier Gastec Corporation™ Supplier No. 12LL

Hydrogen Cyanide Detector Tube for use with Gastec™ Hand Pump.

  • Choose from a variety of detection limits

Specifications

Detection Limits 0.2 to 10 ppm
Components Measured Hydrogen Cyanide
For Use With (Equipment) Gastec Hand Pump
Type Gas Detection Tube
Quantity 1 Tube
Sample Type Gas