missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Chloride - Methanol Reagent (5-10%) [for Esterification], TCI America™

Catalog No. p-7137961
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X004125ML 25 mL
X0041500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X004125ML Supplier TCI America Supplier No. X004125ML
Only null left

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Yellow
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 2924
Chemical Name or Material Hydrogen Chloride - Methanol Reagent (5-10%) [for Esterification]
RTECSNumber MW4025000
TSCA Yes
Recommended Storage Refrigerator