missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hydrogen chloride, 4N solution in 1,4-dioxane, AcroSeal™
CAS: 7647-01-0 | ClH | 36.46 g/mol
$180.41 - $751.51
Specifications
| Chemical Name or Material | Hydrochloric acid |
|---|---|
| Color | Colorless to White |
| Name Note | 4N solution in 1, 4-dioxane |
| CAS | 7647-01-0,123-91-1 |
| Molecular Formula | ClH |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC388361000
|
Thermo Scientific Chemicals
388361000 |
100 mL |
Each for $180.41
|
|
|||||
|
AC388368000
|
Thermo Scientific Chemicals
388368000 |
800 mL |
Each for $751.51
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hydrochloric acid | |
| 4N solution in 1, 4-dioxane | |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| 1.0500g/mL | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 36.45 | |
| 1.05 | |
| Crystals or Crystalline Powder |
| Colorless to White | |
| 7647-01-0,123-91-1 | |
| HCl | |
| 3.95 to 4.40N | |
| 14, 4794 | |
| Solubility in water: soluble. Other solubilities: soluble in most organic solvents | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| AcroSeal™ Glass Bottle | |
| 17°C |
RUO – Research Use Only