missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen bromide, pure, 33 wt.% sol. in glacial acetic acid, AcroSeal™

Catalog No. AC430691000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430691000 100 mL AcroSeal™ Glass bottle
AC430698000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC430691000 Supplier Thermo Scientific Chemicals Supplier No. 430691000

CAS: 10035-10-6 | BrH | 80.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10035-10-6 , 64-19-7
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
SMILES Br

Specifications

Color Yellow
Physical Form Fuming Liquid
Quantity 100 mL
Linear Formula HBr
Flash Point 39°C
Merck Index 11,472
IUPAC Name hydrogen bromide
Formula Weight 80.9
Percent Purity 31 to 34% (HBr)
Grade Pure
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.42
Density 1.4200g/mL
Chemical Name or Material Hydrogen bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 233-113-

RUO – Research Use Only