missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid Regeneration Solution, METTLER TOLEDO™

Catalog No. 01912586
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01912586 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01912586 Supplier METTLER TOLEDO Supplier No. 51350104

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Etches away a very thin layer of glass membranes to expose a fresh surface area

  • Highly diluted
  • Hydrofluoric acid solution
  • Etches away a very thin layer of glass membranes to expose a fresh surface area

Specifications

Chemical Name or Material Hydrofluoric Acid Regeneration Solution
Color Colorless
CAS 1341-49-7
Boiling Point 230™C
Melting Point No Information Available
Molecular Formula (NH4) HF2
Synonym Ammonium Bifluoride, Ammonium Hydrogen Difluoride
Molecular Weight (g/mol) 57.04
Packaging Plastic Bottle
pH ∼2
Quantity 25 mL
Show More Show Less