missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid 47-51%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™

Supplier:  Seastar Chemicals Inc S020502TFIP05

 View more versions of this product

Catalog No. A463-1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1
Colorless
7664-39-3
PFA Bottle
1 L
Liquid
1790
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
F
20.01
CHEBI:29228
OPTIMA
Hydrofluoric Acid
-35°C
105°C
Certificate of Lot included with each bottle.
FH
MFCD00011346
15, 4827
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrogen fluoride
14917
20.01

Chemical Identifiers

7664-39-3
20.01
KRHYYFGTRYWZRS-UHFFFAOYSA-N
14917
hydrogen fluoride
FH
MFCD00011346
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
CHEBI:29228
F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Accessories
Promotions

Promotions